missing translation for 'onlineSavingsMsg'
Learn More

9-Keto Risperidone (90%), TRC

Product Code. 18073730
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073730 10 mg 10mg
17989749 5 mg 5mg
18070230 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073730 Supplier Toronto Research Chemicals Supplier No. TRCK21530010MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189516-65-1
Molecular Formula C23 H25 F N4 O3
Molecular Weight (g/mol) 424.468
Synonym 4H-Pyrido[1,2-a]pyrimidine-4,9(6H)-dione, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-7,8-dihydro-2-methyl-, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-7,8-dihydro-2-methyl-4H-pyrido[1,2-a]pyrimidine-4,9(6H)-dione, Keto paliper, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-7,8-dihydro-4H-pyrido[1,2-a]pyrimidine-4,9(6H)-dione
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-7,8-dihydro-6H-pyrido[1,2-a]pyrimidine-4,9-dione
SMILES CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5CCCC(=O)C5=N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H25FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15H,2-3,6-12H2,1H3
Formula Weight 424.1911
Chemical Name or Material 9-Keto Risperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.