missing translation for 'onlineSavingsMsg'
Learn More

9-Methoxymethyl Guanine, TRC

Product Code. 30478109
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478109 100 mg 100mg
30473142 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478109 Supplier Toronto Research Chemicals Supplier No. TRCM264300100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1202645-50-8
Molecular Formula C7 H9 N5 O2
Molecular Weight (g/mol) 195.18
Synonym 2-Amino-1,9-dihydro-9-(methoxymethyl)-6H-purin-6-one, 9-Methoxymethylguanine, Valganciclovir Hydrochloride Imp. C (EP)
IUPAC Name 2-amino-9-(methoxymethyl)purin-6-ol
SMILES COCn1cnc2c(O)nc(N)nc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H9N5O2/c1-14-3-12-2-9-4-5(12)10-7(8)11-6(4)13/h2H,3H2,1H3,(H3,8,10,11,13)
Formula Weight 195.0756
Chemical Name or Material 9-Methoxymethyl Guanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.