missing translation for 'onlineSavingsMsg'
Learn More

9-Methyladenine, 98%, Thermo Scientific Chemicals

Product Code. 10275481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10275481 100 mg Glass bottle 100mg
10726252 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275481 Supplier Thermo Scientific Acros Supplier No. 325871000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 700-00-5 | Molecular Formula: C6H7N5 | Molecular Weight: 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-00-5
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00047232
InChI Key WRXCXOUDSPTXNX-UHFFFAOYSA-N
Synonym 9-methyladenine, 9-methyl-9h-purin-6-amine, n9-methyladenine, 9-methyl-9h-adenine, 6-amino-9-methylpurine, 9h-purin-6-amine, 9-methyl, adenine, 9-methyl, 9-methyl-9h-purin-6-ylamine, ccris 6905, chembl65976
PubChem CID 69689
ChEBI CHEBI:40526
IUPAC Name 9-methylpurin-6-amine
SMILES CN1C=NC2=C(N)N=CN=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300°C to 302°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 26, V,16, 128
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso
Formula Weight 149.17
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 9-Methyladenine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Methyladenine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.