missing translation for 'onlineSavingsMsg'
Learn More

9-Norketo FK-506, TRC

Product Code. 30417028
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417028 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417028 Supplier Toronto Research Chemicals Supplier No. TRCN6920801MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123719-19-7
Molecular Formula C43H69NO11
Quantity 1 mg
Synonym [3S-[3R*[E(1S*,3S*,4S*)],4S*,5R*,8S*,9E,12R*,14R*,15S*,16R*,18S*,19S*,25aR*]]- 3,4,5,6,8,11,12,13,14,15,16,17,18,19,23,24,25,25a-Octadecahydro-5,19-dihydroxy-3-[2-(4-hydroxy-3-methoxycyclohexyl)-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propenyl)-15,19-epoxypyrido[2,1-c][1,4]oxaazacyclodocosine-1,7,20(22H)-trione
InChI Formula InChI=1S/C43H69NO11/c1-10-13-31-19-25(2)18-26(3)20-37(52-8)40-38(53-9)22-28(5)43(50,55-40)42(49)44-17-12-11-14-32(44)41(48)54-39(29(6)34(46)24-35(31)47)27(4)21-30-15-16-33(45)36(23-30)51-7/h10,19,21,26,28-34,36-40,45-46,50H,1,11-18,20,22-24H2,2-9H3/b25-19+,27-21+/t26-,28+,29+,30-,31+,32-,33+,34-,36-,37-,38+,39+,40+,43+/m0/s1
SMILES C[C@@H]1C[C@@H]([C@@H]2[C@@H](C[C@H]([C@@](O2)(C(=O)N3CCCC[C@H]3C(=O)O[C@@H]([C@@H]([C@H](CC(=O)[C@@H](/C=C(/C1)\C)CC=C)O)C)/C(=C/[C@@H]4CC[C@H]([C@H](C4)OC)O)/C)O)C)OC)OC
Molecular Weight (g/mol) 776.01
Formula Weight 775.4871
Chemical Name or Material 9-Norketo FK-506
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.