missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-2,3,7-trihydroxy-6-fluorone

Product Code. 11470403
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470403 100 g 100g
11450403 5 g 5g
11460403 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470403 Supplier Thermo Scientific Alfa Aesar Supplier No. A11495.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 975-17-7 | C19H12O5 | 320.3 g/mol

9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

Solubility
Soluble in chloroform.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 975-17-7
Molecular Formula C19H12O5
Molecular Weight (g/mol) 320.3
MDL Number MFCD00005048
InChI Key YDCFOUBAMGLLKA-UHFFFAOYSA-N
Synonym phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone
PubChem CID 70420
IUPAC Name 2,6,7-trihydroxy-9-phenylxanthen-3-one
SMILES C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 40910
Solubility Information Soluble in chloroform.
Formula Weight 320.3
Chemical Name or Material 9-Phenyl-2,3,7-trihydroxy-6-fluorone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.