missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylacridine, Thermo Scientific™

Product Code. 17790709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17790709 25 g 25g
17770709 1 g 1g
17780709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17790709 Supplier Thermo Scientific Acros Supplier No. 469430250
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector PHENYLACRIDINE
Chemical Name or Material 9-Phenylacridine
CAS 602-56-2
Melting Point 183°C to 187°C
Color Yellow
Boiling Point 411°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C19 H13 N
MDL Number 00092562
Quantity 25 g
Molecular Weight (g/mol) 255.32
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.