missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole-3-boronic acid, 98%

Product Code. 15421089
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421089 5 g 5g
15431089 250 mg 250mg
15411089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421089 Supplier Thermo Scientific Alfa Aesar Supplier No. H64059.06
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 854952-58-2 | C18H14BNO2 | 287.13 g/mol

9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 854952-58-2
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.13
MDL Number MFCD12196936
InChI Key JWJQEUDGBZMPAX-UHFFFAOYSA-N
Synonym 9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
PubChem CID 51358450
IUPAC Name (9-phenylcarbazol-3-yl)boronic acid
SMILES OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Beilstein 10566519
Solubility Information Slightly soluble in water.
Formula Weight 287.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Phenylcarbazole-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.