Learn More
Uvex™ 9160 I-Vo Replacement Lens
Product Code. 17297241 Shop All Uvex Products
Change view
Click to view available options
Brown
Product Code.
17297241 Brown
1 options
Product Code. 17297241 Supplier Uvex™ Supplier No. 9160118
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Brown tinted polycarbonate replacement lens with supravision sapphire coating.
  • Brown tinted polycarbonate replacement lens
  • Supravision sapphire coating
Supravision Sapphire Coating
  • Inside: Scratch-resistant and chemical-resistant
  • Outside: Scratch-resistant and chemical-resistant
  • Non-stick properties allow for easy removal of greasy/watery dirt
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
Lens Type Supravision Sapphire
Lens Tint Brown
Disposable Reusable
Lens Material Polycarbonate
Labeling 5-2.5