Learn More
uvex i-5 Safety Glasses with CBR65 Lens
Product Code. 17217271 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
17217271 Gray Each
17207271 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17217271 Supplier uvex Supplier No. 9183281
  • ONLINE EXCLUSIVE PRICE
    18.70 €
    16.71 € / Pack of 1
    Save 1.99 €
    10% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reliable protection with an innovative, modern design and the x-tended eyeshield and sideshield. The uvex i-5 can be relied on to protect the eyes in many situations.
  • Sophisticated features ensure superior user comfort when worn for extended periods of time
  • Soft, flexible nosepiece for enhanced comfort and a non-slip fit
  • X-tended eyeshield protects against dust, dirt, and flying particles
  • X-tended sideshield increases protection without restricting peripheral vision
  • 3-position inclination for a personalized fit
  • Extendable x-twist side arm technology offers flexible & convenient adjustment
  • Soft ear pieces for a pressure-free, non-slip fit
  • Reliable UV 400 protection
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 172
Lens Type Supravision Excellence
Lens Coating Anti-fog
Lens Tint Gray
Product Type Ergonomic Safety Glasses
Frame Color Anthracite, Lime
Model 9183 i-5
Transmittance Value 0.23
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-2,5 W 1 FTKN CE