Learn More
uvex 9192 Pheos S Spectacles
Product Code. 17227281 Shop All Uvex Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Clear
Gray
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Lens Tint Unit Size
17227281 Anti-fog, Scratch-resistant Clear Each
17247281 Anti-fog Clear Each
17257281 Anti-fog Gray Each
17237281 Anti-fog, Scratch-resistant Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17227281 Supplier uvex Supplier No. 9192282
  • ONLINE EXCLUSIVE PRICE
    15.10 €
    13.56 € / Pack of 1
    Save 1.54 €
    10% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Duo-spherical lenses provide a wide field of vision with soft, non-slip arm ends to prevent pressure points for enhanced comfort. A narrow version of the original pheo series designed to provide an optimal, secure fit for those with smaller heads.
  • A narrow version of the original pheos series to provide users with smaller heads an optimal, and secure fit
  • Soft, non-slip arm end prevents pressure points for enhanced comfort
  • Duo-spherical lenses provide a large field of vision
  • Design is extremely compatible with ear defenders, helmets, and bump caps
  • Eye Plus Protection provides additional eyebrow protection with a bevelled edge in the upper area
  • Easily add a headband and frame when working at heights or in confined spaces
Supravision Extreme Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
Lens Type Duo-Spherical
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Ergonomic Safety Glasses
For Use With (Application) Working at Height or in Confined Spaces
Frame Color Black, Gray
Model Pheos S
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Narrow
Labeling W 166 F CE - 2C-1.2 W 1 FKN CE