Learn More
uvex 9192 Pheos Safety Glasses
Product Code. 15316082 Shop All Uvex Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Orange
Blue, Gray
Lens Tint:
Clear
Gray
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color Lens Tint Unit Size
15316082 Black, Gray Gray Each
15386072 Black, Orange Gray Each
15386082 Blue, Gray Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15316082 Supplier uvex Supplier No. 9192285
  • ONLINE EXCLUSIVE PRICE
    13.90 €
    13.65 € / Pack of 1
    Save 0.25 €
    1% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pheos safety glasses feature a large field of vision thanks to the duosphere disc, and excellent ventilation to prevent fogging.
  • Perfect all-round protection, with a large field of vision through duo spherical lens
  • Outstanding ventilation thanks to integrated uvex climazone temple concept
  • Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical-resistant on the outside
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Supravision Excellence
Lens Coating Anti-fog
Certifications/Compliance (Europe) EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Tint Gray
Product Type Ergonomic Safety Glasses
Frame Color Black, Gray
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard