Learn More
uvex 9194 I-Works Safety Spectacles
Product Code. 17277291 Shop All Uvex Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Silver Mirror
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Lens Tint unitSize
17277291 Anti-fog Silver Mirror Each
17267291 Anti-fog, Scratch-resistant Amber Each
17247291 Anti-fog, Scratch-resistant Clear Each
17257291 Anti-fog, Scratch-resistant Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17277291 Supplier uvex Supplier No. 9194885
Only null left
Add to basket
Add to basket
Spectacles feature a light, sporty design with panoramic lenses to ensure maximum comfort even when worn for long periods.
  • Spectacles combine an attractive appearance with robust quality
  • Light, sporty design with panoramic lenses ensure maximum comfort
  • Permanent anti-fog performance, even after repeated cleaning
  • Soft, duo-component arms for a comfortable non-slip grip
  • Flat-end arm design with opening to attach a neck cord
  • Available with a sunglare filter (9194885)
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Type Panoramic
Lens Coating Anti-fog
Filter Type Sunglare
Lens Tint Silver Mirror
Product Type Ergonomic Safety Glasses
Frame Color Black, Gray
Model 9194 i-Works
Transmittance Value 0.53
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-1.7 W 1 FTN CE