Learn More
uvex 9301 Ultravision Gas-Tight Goggles
Product Code. 17217321 Shop All Uvex Products
Change view
Click to view available options
Red
Product Code.
17217321 Red
1 options
Product Code. 17217321 Supplier uvex Supplier No. 9301633
  • ONLINE EXCLUSIVE PRICE
    20.60 €
    18.52 € / Pack of 1
    Save 2.08 €
    10% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ergonomic, full-view goggles are gas-tight and feature supravision excellence coating, with a heat-resistant support body material.
  • Comfortable, ergonomically designed full-view goggles
  • Suitable for glasses wearers, unrestricted side visibility, problem-free lens change
  • Clear, polycarbonate lens with supravision excellence coating
  • Gas-tight without a ventilation system
  • Heat resistant support body material
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
  • Easy to clean, and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
Lens Type Supravision Excellence
Lens Coating Anti-fog
Description Gastight
Color Red
Lens Tint Clear
Product Type Anti-fog Goggles
Model 9301 Ultravision
Ventilation Details Not Ventilated
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 345 BT CE - 2C-1,2, W1 BTKN CE