missing translation for 'onlineSavingsMsg'
Learn More

uvex 95798 Insole For Uvex 1 Sport - Width 11

Product Code. 17298981 Shop All Uvex Products
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
51
Unit Size:
1 pair
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17298981 51 1 pair
17248971 35 1 pair
17258971 36 1 pair
17268971 37 1 pair
17278971 38 1 pair
17288971 39 1 pair
17298971 40 1 pair
17208981 41 1 pair
17218981 42 1 pair
17228981 43 1 pair
17238981 44 1 pair
17248981 45 1 pair
17258981 46 1 pair
17268981 47 1 pair
17278981 48 1 pair
17288981 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 17298981 Supplier uvex Supplier No. 9579851
  • ONLINE EXCLUSIVE PRICE
    20.10 €
    18.15 € / 1 pairs
    Save 1.95 €
    9% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Comfortable climatic insole with antistatic properties for Uvex 1 Sport

  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9579.8
  • Sizes: 35 to 52
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Uvex 1 Sport
Material Microvelour
Size 51
Disposable Single-use
Gender Unisex
Fit Standard Width
Latex Free Yes
Waterproof No
Product Type Insoles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.