missing translation for 'onlineSavingsMsg'
Learn More

Uvex 95980 Insole For Uvex Naturform - Width 11

Product Code. 17219021 Shop All Uvex Products
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17219021 38 1 pair
17229021 39 1 pair
17239021 40 1 pair
17249021 41 1 pair
17259021 42 1 pair
17269021 43 1 pair
17279021 44 1 pair
17289021 45 1 pair
17299021 46 1 pair
17209031 47 1 pair
17219031 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 17219021 Supplier Uvex Supplier No. 9598038

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Comfortable climatic insole with antistatic properties for Uvex Performance (Naturform)

  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9598.0
  • Sizes: 38 to 48
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Uvex Naturform
Material Microvelour
Size 38
Disposable Reusable
Gender Unisex
Fit Standard width
Latex Free Yes
Waterproof No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.