missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3730xl DNA Analyzer 96-Capillary Array, 36 cm

Product Code. 15869766 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869766 1 array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869766 Supplier Applied Biosystems™ Supplier No. 4331244
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each array contains 96 capillaries (36cm in length) for use with the Applied Biosystems 3730xl DNA analyzer.

This is a capillary array for the Applied Biosystems 3730xl DNA analyzer. Each array contains 96 capillaries, 36cm long.

Used for sequencing, microsatellite, SNP genotyping, and fragment analysis runs.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Sequencing
Software Type Fragment Analysis
Quantity 1 array
For Use With (Equipment) 3730xl DNA Analyzer
Length (Metric) 36 cm (Capillary)
Number of Capillaries 96 Capillaries
Product Type Capillary Array
Capillary Length 36 cm
Shipping Condition Room Temperature
Content And Storage Each array contains 96 capillaries (36 cm in length) for use with the Applied Biosystems 3730xl DNA Analyzer.

We guarantee the performance of the 3730 Capillary arrays up to 300 runs and within 1 year of purchase.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.