Learn More
Eppendorf™ 96-channel Dispensing Heads
Product Code. 30040081 Shop All Eppendorf Products
Change view
Click to view available options
Device Holds:
0.5 to 300 μL
5 to 1000 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Device Holds Unit Size
30040081 0.5 to 300 μL Each
30040079 5 to 1000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30040081 Supplier Eppendorf™ Supplier No. 5069000527
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with epMotion™ 96 Flex in hard-shell case. Requires base device for operation.
  • For use with epMotion™ 96 Flex (not included)
  • Intuitive semi-automated handling of 96- and 384-well plates
  • Calibrated according to ISO 23783-2 standards
  • Includes hard shell case for transport and storage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dispensing Head
Autoclavable Not Autoclavable
Device Holds 0.5 to 300 μL
For Use With (Application) Cell culture, Molecular biology
For Use With (Equipment) epMotion™ 96 Flex, and epT.I.P.S.™ Motion Reload System
Quantity 1 Ea.
Weight (English) 4.6 lb.
Weight (Metric) 2.1 kg
No. per Pack 1 Ea.