Learn More
BTX™ 96-Well Disposable Electroporation Plate
Product Code. 15721078
Change view
Click to view available options
Pulse Form:
Exponential Decay Wave, Square Wave
Unit Size:
Each
Product Code. Pulse Form Unit Size
15721078 Exponential Decay Wave, Square Wave Each
1 options
Product Code. 15721078 Supplier BTX™ Supplier No. 450450M
  • ONLINE EXCLUSIVE PRICE
    346.00 €
    294.92 € / Pack of 1
    Save 51.08 €
    14% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
96-Well Disposable Electroporation Plate are sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Try out many different electroporation conditions (cell density, buffer, pulse voltage and width, etc.) to find the highest transfection efficiency, quickly and easily with the ECM 830HT, ECM630 HT or the Gemini X2 HT Electroporation systems.
  • Standard multi-well formats
  • Bio-compatible materials
It's like 96 electroporation cuvettes in one convenient plate.
Specifications
For Use With (Application) High-throughput Electroporation
For Use With (Equipment) BTX High-throughput Systems
Description 96-well Disposable Electroporation Plate, 2 mm gap, 100 μL, 1 Plate
Quantity 1 Ea.
Size (English) Gap 0.08 in.
Size (Metric) Gap 2 mm
Volume (Metric) 100 μL
Sterility Sterile
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes