missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Plate Septa, 96 well

Product Code. 12059570 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 each
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12059570 20 each Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12059570 Supplier Applied Biosystems™ Supplier No. 4315933
Black Friday Starts Here
Use promo code 27984
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each package contains 20 septa, sufficient for 1,920 samples

These are septa for use with the 3730, 3730xl, 3130xl, and 3130, instruments.

These are septa for use with the 3730, 3730xl, 3130xl, and 3130, instruments.

Use with:
310 Running Buffer (10X), 3100/3100-Avant™ Genetic Analyzer Autosampler Plate Kit, 96-well, 3130/3100-Avant™ Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (36 cm), 3130xl/3100 Genetic Analyzer Capillary Array (50 cm), 3730 Running Buffer (10X), 5.0 mL Glass Syringe (polymer reserve syringe), 96-Well Plate Base, 96-Well Plate Retainer, Hi-Di™ Formamide, MicroAmp™ Optical 96-Well Reaction Plate, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, POP-4™ Polymer for 3100/3100-Avant™ Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, POP-7™ Polymer for 3130/3130xl Genetic Analyzers, Reservoir Septa

TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 each
Format 96-well Plate
For Use With (Equipment) 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3730 DNA Analyzer, 3130 Genetic Analyzer
Product Type Septa Plate
Shipping Condition Room Temperature
Content And Storage Each package contains 20 septa, sufficient for 1,920 samples.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.