missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well tips, Clear
BIOPHARMA

Product Code. 16620234 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
200 μL
250 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Tip Style:
Low Retention, Maxymum Recovery
Standard
Unit Size:
Pack of 4800
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Tip Style unitSize
16620234 250 μL Sterile Non-filtered Standard Pack of 4800
16640234 200 μL Sterile Filtered Standard Pack of 4800
16630234 200 μL Sterile Filtered Low Retention, Maxymum Recovery Pack of 4800
16600234 250 μL Non-sterile Non-filtered Low Retention, Maxymum Recovery Pack of 4800
16610234 250 μL Sterile Non-filtered Low Retention, Maxymum Recovery Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16620234 Supplier Axygen™ Supplier No. VT255R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Axygen™ Maxymum Recovery™ tips have been developed to work with the Agilent/Velocity11 and MGI Tech automation systems, which can be enabled for the high throughput of magnetic bead based RNA extraction of biological samples or pre-configured and qualified to automate high throughput pre-PCR samples.

  • Tip format: 96 tips
  • Tip volume range: Up to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    • With or without aerosol-resistant filters
    • non-sterile or sterile
    • Axygen Maxymum Recovery surface
  • Compatibility:
    • Bravo automated liquid handling platform
    • VPrep™ vertical liquid handling pipetting station
    • MGISP-960
    • MGISP-100
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Format Rack, 96
Tip Style Standard
Quantity 96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs
For Use With (Equipment) Agilent Bravo and MGI Tech MGISP Automation Systems, VPrep Vertical Liquid Handling Pipetting Station
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.