missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ 96-Well Twin.tec™ PCR Plates, Non-skirted
Non-skirted 96-Well Twin.tec™ PCR Plates are ideal for extreme applications such as multiple heat sealing.
Brand: Eppendorf™ 0030133366
Code : 96
Additional Details : Weight : 0.66700kg
Description
- Fits even more thermal cyclers
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors
- Available with regular (250 μL) or low profile (150 μL) wells
- Ideal for quantitative real-time PCR as well
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
Specifications
No Barcode | |
Standard | |
Autoclavable | |
PCR Plate 96 | |
Polycarbonate | |
96 | |
Regular | |
Colorless |
RNase, DNase free | |
8 x 12 | |
Clear | |
Non-skirt | |
250 μL | |
Ideal for extreme applications such as multiple heat sealing. | |
PCR Clean |