missing translation for 'onlineSavingsMsg'
Learn More

uvex 96983 Shoe Spray

Product Code. 17209061 Shop All Uvex Products
Change view
Click to view available options
Waterproof:
No
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Waterproof Unit Size
17209061 No Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17209061 Supplier uvex Supplier No. 9698300
  • ONLINE EXCLUSIVE PRICE
    21.00 €
    17.15 € / Pack of 1
    Save 3.85 €
    18% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Uvex Nano shoe footwear disinfectant, against foot odor and athlete's foot

  • Uvex Nano Shoe uses specially developed technology which releases controlled doses of active ingredients with long-lasting effect
  • Disinfectant contains antimicrobial and antifungal active ingredients
    • Eliminate bacteria and fungus long term
  • Formula contains only active ingredients approved for use for number of years, found in cosmetic (formaldehyde-free, phenol-free)
  • Odor prevention:
    • Regular application in new shoes prevents any buildup of odor from outset
    • Deals with bacteria
  • Ongoing deodorization:
    • Returns footwear that has already been well-worn to odor neutral conditions
  • Kills fungi
  • For use on inside surfaces of all footwear
  • Article No. 9698.300
  • Contains 125 mL

Use:

  • Spray in Uvex Nano Shoe about 5 to 10 doses into footwear outside in open air
  • After 5 min. drying, shoes are ready to wear
  • Recommended initial treatment of badly affected footwear 3 applications/week
  • Regular use prevents odors and fungus developing long term
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Uvex Nano
Disposable Reusable
Gender Unisex
Resistant To Static
Latex Free Yes
Waterproof No
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.