missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9790 Visor Adaptors

Product Code. 17259201 Shop All Uvex Products
Change view
Click to view available options
For Use With (Equipment):
Pheos Alpine
Pheos K2H Magnet (2600205)
Unit Size:
1 pair
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17259201 Pheos Alpine Each
17239211 Pheos K2H Magnet (2600205) 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17259201 Supplier Uvex™ Supplier No. 9790067
  • ONLINE EXCLUSIVE PRICE
    8.60 €
    8.42 € / Pack of 1
    Save 0.18 €
    2% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For attaching visors to safety helmets.

  • Adapter for attaching visors to Uvex™ safety helmets
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Pheos Alpine
For Use With (Application) Attaching Visors to Safety Helmets
Product Type Visor Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.