missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9906 Pheos Polycarbonate Visors

Product Code. 17260272 Shop All Uvex Products
Change view
Click to view available options
Mounting Type:
Magnetic Locks
Mechanical Levers
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Type Unit Size
17260272 Magnetic Locks Each
17250272 Mechanical Levers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17260272 Supplier Uvex™ Supplier No. 9906003
  • ONLINE EXCLUSIVE PRICE
    73.90 €
    66.11 € / Pack of 1
    Save 7.79 €
    10% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Visors are treated with supravision excellence coating for anti-fog and anti-scratch protection. Available with two alternative mountings: mechanical levers or automatic magnetic hooks.

  • Magnetic locking system is extremely easy to attach to the visor
  • Ergonomic joint system is comfortable to wear, even for long periods
  • Compatible with Pheos B-S-WR and Pheos Alpine (Adapter 9790067 is required for combination with Alpine)
  • Visor 9906002 can be combined with earmuffs Pheos K2H Earmuffs (Mechanical)
  • Visor 9906003 is compatible with Pheos K2H Earmuffs (Magnetic)
  • Certified for areas of application 3 (liquids) and 9 (molten metal/hot solids)
  • Labeling: W 166 3 8-2 9 B CE 0196 (Frame), 2C-1.2 W 1 B 9 KN CE 0196 (Lens)
  • Lens can be changed
  • Helmet sold separately

Supravision Excellence Coating

  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 166:2001, EN 170:2002
Color Black
For Use With (Equipment) Pheos B-S-WR, Pheos Alpine (Requires Adaptor 9790067), and Pheos K2H Earmuffs (Magnetic)
Lens Tint Clear
Material Polycarbonate
Model 9906 Pheos
Mounting Type Magnetic Locks
Product Type Visor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.