missing translation for 'onlineSavingsMsg'
Learn More

uvex 9972 Cleaning Fluid

Product Code. 17250292 Shop All Uvex Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17250292 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17250292 Supplier uvex Supplier No. 9972101
  • ONLINE EXCLUSIVE PRICE
    11.60 €
    11.45 € / Pack of 1
    Save 0.15 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An effective cleanser for use with all Uvex™ Safety Spectacles.

  • Packaged in a 500 mL round bottle
  • Suitable for use with all viewing lenses
  • Not compatible with Cleaning Station 9970005
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Product Type Cleaning Fluid
For Use With (Equipment) Uvex™ Viewing Lenses
Quantity 500 mL
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.