missing translation for 'onlineSavingsMsg'
Learn More

9H-Carbazole, TRC

Product Code. 17970735
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970735 100 mg 100mg
17974205 10 g 10g
17952405 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970735 Supplier Toronto Research Chemicals Supplier No. TRCC176065100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12 H9 N
Molecular Weight (g/mol) 167.21
Synonym Carbazole (8CI), 9-Azafluorene, Chlorophenesin carbamate, Dibenzo[b,d]pyrrole, Dibenzopyrrole, Diphenylenimine, NSC 3498, SKF 20091
IUPAC Name 9H-carbazole
SMILES c1ccc2c(c1)[nH]c3ccccc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H
Formula Weight 167.0735
Chemical Name or Material Carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.