missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 9mm Certified Amber Wide Opening Screw Thread Kits

Product Code. 15290379
Change view
Click to view available options
Description:
2mL Amber I-D; Black Cap; Bonded PTFE/Silicone Septum
2mL; Amber ID Vial; With 9mm Preslit PTFE/Silicone Septa/Cap
Assembled Kit, 2mL Certified 9mm Wide Opening Screw Vial
Certified Target DP 2mL Amber Glass I-D Vial, 9mm Screw Thread
Certified Target DP 2mL Amber Glass I-D, 9mm Screw Thread
Convenience Kit, 2mL 9mm Wide Opening Screw Vial
Convenience Kit, 2mL 9mm Wide Opening Screw Vial, Silanized
Convenience Kit, 2mL Certified 9mm Wide Opening Screw Vial
Convenience Kit, 2mL Certified 9mm Wide Opening SureStop Vial
Closure Color:
Black
Blue
Gray
Septum:
Blue PTFE/White Silicone
Bonded Clear PTFE/Clear Silicone
Bonded PTFE/Silicone
Bonded Red PTFE/ White Silicone
Bonded Red PTFE/White Silicone
Ivory PTFE/Red Rubber
PTFE/Silica
PTFE/Silicone
Red PTFE/White Silicone
Unit Size:
Pack of 100
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Closure Color Septum unitSize
15290379 Convenience Kit, 2mL Certified 9mm Wide Opening SureStop Vial Blue Blue PTFE/White Silicone Pack of 100
11951543 Certified Target DP 2mL Amber Glass I-D, 9mm Screw Thread - PTFE/Silicone Pack of 100
10240665 2mL Amber I-D; Black Cap; Bonded PTFE/Silicone Septum Black Bonded PTFE/Silicone Pack of 100
10169464 2mL; Amber ID Vial; With 9mm Preslit PTFE/Silicone Septa/Cap Gray Bonded Red PTFE/White Silicone Pack of 100
15220389 Convenience Kit, 2mL Certified 9mm Wide Opening SureStop Vial Blue Ivory PTFE/Red Rubber Pack of 100
15280379 Convenience Kit, 2mL Certified 9mm Wide Opening SureStop Vial Blue Red PTFE/White Silicone Pack of 100
15260389 Convenience Kit, 2mL Certified 9mm Wide Opening Screw Vial Blue Ivory PTFE/Red Rubber Pack of 100
11951513 Certified Target DP 2mL Amber Glass I-D Vial, 9mm Screw Thread - PTFE/Silica Pack of 100
15318066 Convenience Kit, 2mL 9mm Wide Opening Screw Vial Blue Bonded Clear PTFE/Clear Silicone Pack of 100
15368066 Convenience Kit, 2mL 9mm Wide Opening Screw Vial Gray Bonded Clear PTFE/Clear Silicone Pack of 100
15398066 Convenience Kit, 2mL 9mm Wide Opening Screw Vial, Silanized Gray Bonded Clear PTFE/Clear Silicone Pack of 100
15388036 Assembled Kit, 2mL Certified 9mm Wide Opening Screw Vial Black Bonded Red PTFE/White Silicone Pack of 100
15398036 Assembled Kit, 2mL Certified 9mm Wide Opening Screw Vial Gray Bonded Red PTFE/ White Silicone Pack of 100
15358046 Convenience Kit, 2mL Certified 9mm Wide Opening Screw Vial Black Bonded Red PTFE/ White Silicone Pack of 100
15364654 Convenience Kit, 2mL Certified 9mm Wide Opening Screw Vial Gray Bonded Clear PTFE/Clear Silicone Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 15290379 Supplier Thermo Scientific™ Supplier No. CERT5000576W

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.

Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ 9mm Wide Opening Amber Glass Screw MS Certified Vial Convenience Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.

Unassembled kits

  • 100 vials and 100 caps with pre-assembled septa 
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together 
  • Clear trays make it easy to keep track of available supplies without opening containers

Assembled kits

  • 100 vials with pre-attached caps and septa 
  • Packaged in convenient vial trays with clear covers
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Description Convenience Kit, 2mL Certified 9mm Wide Opening SureStop Vial
Color Amber
Material Glass, PTFE, Silicone
Pre-slit Yes
Septum Blue PTFE/White Silicone
No. per Pack 100
Product Line LC-GC CERT
Marking Spot Yes

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.