missing translation for 'onlineSavingsMsg'
Learn More

(9S,12R,13S)-Pinellic Acid, TRC

Product Code. 30410118
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410118 10 mg -
30416504 5 mg -
30411277 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410118 Supplier Toronto Research Chemicals Supplier No. TRCP10273510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102735-58-0
Molecular Formula C18H34O5
Molecular Weight (g/mol) 330.47
Synonym (9S,10E,12R,13S)-9,12,13-Trihydroxy-10-octadecenoic Acid, [9S-(9R*,10E,12S*,13R*)]-9,12,13-Trihydroxy-10-Octadecenoic Acid
IUPAC Name (E,9S,12R,13S)-9,12,13-trihydroxyoctadec-10-enoic acid
SMILES CCCCC[C@H](O)[C@H](O)\C=C\[C@@H](O)CCCCCCCC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H34O5/c1-2-3-7-11-16(20)17(21)14-13-15(19)10-8-5-4-6-9-12-18(22)23/h13-17,19-21H,2-12H2,1H3,(H,22,23)/b14-13+/t15-,16-,17+/m0/s1
Formula Weight 330.2406
Chemical Name or Material (9S,12R,13S)-Pinellic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.