missing translation for 'onlineSavingsMsg'
Learn More

(9Z)-9-Octadecen-1-ol, 85%, TRC

Product Code. 17944845
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944845 50 g 50g
17983095 100 g 100g
17964755 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944845 Supplier Toronto Research Chemicals Supplier No. TRCO23593550G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18 H36 O
Molecular Weight (g/mol) 268.48
Synonym Oleyl Alcohol
IUPAC Name octadec-9-en-1-ol
SMILES CCCCCCCCC=CCCCCCCCCO

Specifications

Quantity 50 g
InChI Formula InChI=1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3
Formula Weight 268.2766
Chemical Name or Material Oleyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.