Learn More
Gilson™ Maintenance Kit for Pipetman™
Manufactured by Gilson™. Gilson™ Maintenance Kit for Pipetman™ are guaranteed to deliver value and assure reliability and precision.
Brand:  Gilson™ F144501
Product Code. 10133192
  • ONLINE EXCLUSIVE PRICE
    83.45 €
    81.17 € / Each
    Save 2.28 €
    2% Off
Description
  • Maintenance kit  
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For Pipetman: P2/P2G
1 Tip-holder, 3 piston seals and 3 O-rings