missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, HPLC Grade, 99% min, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 022932.K7

Product Code. 11388207

  • 259.00 € / 4 L
 Restricted Item
Estimated Shipment: 21-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

71-23-8
97°C
99% min.
C3H8O
MFCD00002941
1274
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
HPLC
0.804 g/cm3 at 20°C
15°C (59°F)
Alcohol-like
1.385
4L
1098242
Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
CCCO
60.10
CHEBI:28831
13.3 hPa (10mm Hg) at 20°C
1-Propanol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.