missing translation for 'onlineSavingsMsg'
Learn More

Nigericin sodium salt, 98+%, Thermo Scientific™

A potassium ionophore

Brand:  Thermo Scientific Alfa Aesar J61349.MA

206.85 EUR valid until 2026-09-25
BEST PRICE on Promo! Use promo code "27966" to get your promotional price.



Quantity 10mg

Product Code. 15403959

  • 209.00 € / 10mg
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Nigericin sodium salt is a polyether ionophore which catalyzes the electroneutral exchange of alkali metal (K+) for H+ (antiport). It disrupts membrane potential and stimulates ATPase activity in mitochondria. It transports monovalent cations across membranes with the following specificity: K+ >Rb+ >Cs+ >>Na. It kills bacteria by facilitating the diffusion of ions across membranes. It is widely used in studies of the consequences of changes in membrane potential in variable systems.

Solubility
Soluble in chloroform, methanol or ethanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, bases and heat.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

28643-80-3
MFCD00036825
UN3462
14,6541
Soluble in chloroform, methanol or ethanol
CC1CCC(OC1C(C)C(=O)O)CC2CC(C(C3(O2)C(CC(O3)(C)C4CCC(O4)(C)C5C(CC(O5)C6C(CC(C(O6)(CO)O)C)C)C)C)C)OC
724.973
746.94
Nigericin sodium salt
C40H68O11
10mg
3892398
nigericin
DANUORFCFTYTSZ-FBMOGUJMSA-N
(2R)-2-[(2R,3S,6R)-6-[[(2S,4R,5R,6R,7R,9R)-2-[(2R,5S)-5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-
123134611
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nigericin sodium salt

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Nigericin sodium salt

SDS Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-09-25
Curiosity Goes Beyond
Promo Code: 27966
Push the boundaries of discovery with exceptional savings across essential research workflows: Cell Culture and Analysis driven by our Gibco™ product portfolio, Gene Expression powered by Applied Biosystems™ solutions, and Protein Expression enabled by Invitrogen™ and Thermo Scientific™ products.

Applying Promo and Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.