missing translation for 'onlineSavingsMsg'
Learn More

Bacitracin, Thermo Scientific™

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.

Brand:  Thermo Scientific Alfa Aesar J62432.06

50.33 EUR valid until 2026-03-27
BEST PRICE on Promo! Use promo code "27964" to get your promotional price.



Quantity 5g

Product Code. 15404819

  • 71.90 € / 5g
Curiosity Meets Adventure
Use promo code 27964
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945
• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components
Applications
• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity
• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1405-87-4
5g
MFCD00062640
Soluble in water.
Bacitracin
White
C66H103N17O16S
14,934
1421.6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bacitracin

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contain: Bacitracin

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.