missing translation for 'onlineSavingsMsg'
Learn More

Anisomycin, 97%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62964.MF

Quantity 50mg

Product Code. 15415259

  • ONLINE EXCLUSIVE PRICE
    274.00 €
    269.00 € / 50mg
    Save 5.00 €
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A protein synthesis inhibitor
  • Causes apoptosis of PC12 Cells
TRUSTED_SUSTAINABILITY
Specifications

Specifications

22862-76-6
50mg
MFCD00077650
20705
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin
YKJYKKNCCRKFSL-BFHYXJOUSA-N
(2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
253602
265.31
Anisomycin
White
C14H19NO4
UN3462
14,670
Soluble in water at 2mg/ml,in DMSO at 20mg/ml,and in methanol at 20mg/ml
COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1
265.31
CHEBI:338412
97%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.