missing translation for 'onlineSavingsMsg'
Learn More

Suramin hexasodium salt, 98+%, Thermo Scientific™

Suramin hexasodium salt (suramin sodium, CAS # 129-46-4) is the sodium salt of suramin.

Brand:  Thermo Scientific Alfa Aesar J61707.MD

Quantity 250mg

Product Code. 15454249

  • 356.00 € / 250mg
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Suramin is a polysulphonated napthylurea which exhibits anti-parasitic and anticancer properties

• Suramin prevents endothelial cell migration and proliferation through inhibition of IGF-I, EGF, PDGF, and TGF-beta

• Suramin can also uncouple G-proteins from receptors and turn off cell signaling cascades

Application

• Suramin inhibits angiogenesis through blockade of bFGF and VEGF and can be useful for the laboratory study of blood vessel growth and renewal following injury

• Suramin inhibits DNA topoisomerases, and can be used as an antineoplastic agent in the laboratory study of cancer or for studies of microbial physiology

TRUSTED_SUSTAINABILITY
Specifications

Specifications

129-46-4
MFCD00210217
3694087
suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
VAPNKLKDKUDFHK-UHFFFAOYSA-H
hexasodium 8-{4-methyl-3-[3-({[3-({2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonate
8514
≥98%
C51H34N6Na6O23S6
250mg
14,9006
Soluble in water and saline; Sparingly soluble in 95% bethanol
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
1429.15
1429.16
Suramin hexasodium salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.