missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, Thermo Scientific™

Nystatin, CAS # 1400-61-9, is a polyene compound that shows anti-bacterial and antifungal activities. It is also an ionophore that increases the membrane permeability to some ions.

Brand:  Thermo Scientific Alfa Aesar J62486.09

Quantity 10g

Product Code. 15454869

  • 122.00 € / 10g
 Restricted Item
In Stock
Edge
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Nystatin is a polyene compound that shows anti-bacterial and antifungal activities against Streptomyces noursei, Streptomyces aureus, and other Streptomyces species
• It is also an ionophore with the capacity to increase membrane permeability
• Nystatin is a channel-forming ionophore and exerts its activity via formation of a membrane-spanning pore in the fungal plasma membrane, thus altering membrane permeability, allowing the leakage of intracellular contents, and the subsequent disruption of electrochemical gradients essential for proper cell function
Applications
• In in vitro experiments, Nystatin is an antifungal that is both fungistatic and fungicidal against a wide variety of yeasts and yeast-like fungi
• It has a higher selectivity for fungal cells over mammalian cells since nystatin presents greater binding affinity with ergosterol, a key sterol found in fungal cell walls, as opposed to its mammalian counterpart, cholesterol
• This compound is not active against organisms that do not contain sterols in their cell membrane (for example, bacteria)
• It is used on human macrophage cell line U937, to study its effect on the production of interleukin-8 by silver nanoparticle-treated U937 cell line
• It has the ability to prevent cell culture contamination as well as induce interleukin-1 and -8, and tumor necrosis factor α secretion in TLR2-expressing THP1 cells
• It modulates lipid raft profile in mice macrophages
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1400-61-9
MFCD00036240
14,6736
VQOXZBDYSJBXMA-AWCIUYGFSA-N
(1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
133640190
Nystatin
C47H75NO17
10g
Soluble in DMSO
CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
926.107
926.09
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.