missing translation for 'onlineSavingsMsg'
Learn More

Phthalan, 96%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 045592.06

Quantity 5g

Product Code. 15472387

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

496-14-0
192°C
C8H8O
MFCD00005932
111932
SFLGSKRGOWRGBR-UHFFFAOYSA-N
1,3-dihydro-2-benzofuran
10327
96%
1.094 g/mL
63°C (145°F)
1.546
5g
phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
C1C2=CC=CC=C2CO1
120.151
120.15
Phthalan
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.