missing translation for 'onlineSavingsMsg'
Learn More

Aluminum iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047170.14

Product Code. 15477435

  • 622.20 € / 25g
 Restricted Item
Estimated Shipment: 21-12-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Specifications

Specifications

7784-23-8
AlI3
UN3260
aluminum iodide, aluminium iodide, aluminium triiodide, aluminium iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
[Al](I)(I)I
407.695
407.69
Pungent
Moisture & Light Sensitive
(metals basis)
25g
MFCD00003427
14,346
CECABOMBVQNBEC-UHFFFAOYSA-K
triiodoalumane
82222
99.999%
Ampule
3.98g/mL
Aluminum iodide, Ultra dry
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.