missing translation for 'onlineSavingsMsg'
Learn More

IKA™ A 11 Basic
GREENER_CHOICE

Product Code. 16208111 Shop All IKA Products
Change view
Click to view available options
Plug Type:
UK plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
16208111 UK plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16208111 Supplier IKA™ Supplier No. 0002900002
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Batch mill with high-grade stainless steel beater is designed for impact grinding of hard, brittle, or non-elastic grinding materials.

  • Moist and gluey materials can be pulverized by adding water
  • Grinding chamber made of PTFE, glass fiber-reinforced with stainless steel inlet (1.4571)
  • Useful volume of 80 mL
  • For embrittlement of grinding materials with liquid nitrogen in the grinding chamber
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W x H) 85 x 85 x 240 mm
Hertz 50/60 Hz
Plug Type UK plug
Speed 0 to 28 rpm
Voltage 100/115 V, 220/230 V
Weight (Metric) 1.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.