missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 196

Description
A 196 is a potent and selective SUV420H1/H2 inhibitor (IC50 values are 25 and 144 nM respectively) that exhibits 100-fold selectivity over other histone methyltransferases and non-epigenetic targets. A 196 inhibits the di- and tri-methylation of H4K20me in multiple cell lines (IC50 < 1 μM).
To request the negative control for A 196, please fill out the SGC 2043 request form on the SGC website.
Specifications
Specifications
| Chemical Name or Material | 6,7-Dichloro-N-cyclopentyl-4-(4-pyridinyl)-1-phthalazinamine |
| CAS | 1982372-88-2 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C18H16Cl2N4 |
| Formula Weight | Observed MW: 359.25 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?