missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 205804
GREENER_CHOICE

Product Code. 18074052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074052 10 mg 10mg
18065752 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074052 Supplier Tocris Bioscience™ Supplier No. 2524/10
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective inhibitor of E-selectin and ICAM-1 expression

Selective inhibitor of E-selectin and ICAM-1 expression (IC50 values are 20, 25 and > 1000 nM for TNF-α-induced E-selectin, ICAM-1 and VCAM-1 expression respectively). Effective inhibitor of cell-cell adhesion in an in vitro flow experiment.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251992-66-2
Molecular Formula C15H12N2OS2

Specifications

Quantity 10 mg
Formula Weight 300.39
Purity >98%
Molecular Weight (g/mol) 300.394
Chemical Name or Material A 205804
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.