missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 286982
GREENER_CHOICE

Product Code. 18094222
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18094222

Brand: Tocris Bioscience™ 4228/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent inhibitor of the LFA-1/ICAM-1 interaction

Potent inhibitor of the LFA-1/ICAM-1 interaction (IC50 values are 44 and 35 nM in LFA-1/ICAM-1 binding and LFA-1-mediated cell adhesion assays respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280749-17-9
Molecular Formula C24H27N3O4S

Specifications

Quantity 10 mg
Formula Weight 453.55
Purity >99%
Molecular Weight (g/mol) 453.557
Chemical Name or Material A 286982
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.