missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 33
GREENER_CHOICE

Product Code. 18796036
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18796036

Brand: Tocris Bioscience™ 6313/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PDE4B inhibitor

A 33 is a potent and selective PDE4B inhibitor (IC50 = 15 nM); exhibits >100-fold selectivity for PDE4B over 4D. Regulates cAMP in inflammatory cells. Inhibits LPS-induced TNF-α production and neutrophil accumulation in mouse lungs. Reduces microglial and neutrophil activation, and improves behavioral deficits in a traumatic brain injury model in vivo. Also exhibits antidepressant-like effects. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-[[2-(5-Chloro-2-thienyl)-5-ethyl-6-methyl-pyrimidin-4-yl]amino]phenyl]acetic acid
CAS 915082-52-9
Quantity 10 mg
Molecular Formula C19H18ClN3O2S
Formula Weight 387.88
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.