missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 419259 trihydrochloride

Description
Potent Hck inhibitor (IC50 = 0.43 nM). Also inhibits other Src family kinases (SFKs). Suppresses proliferation and induces apoptosis in CML cell lines including CD34+. Also inhibits acute myeloid leukemia (AML) stem cell proliferation in vitro and in vivo.
Specifications
Specifications
| CAS | 1435934-25-0 |
| Quantity | 10 mg |
| Molecular Formula | C29H37Cl3N6O |
| Formula Weight | 592.01 |
| Purity | >98% |
| Molecular Weight (g/mol) | 592.006 |
| Chemical Name or Material | A 419259 trihydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?