missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 425619
GREENER_CHOICE

Product Code. 18782148
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18782148

Brand: Tocris Bioscience™ 5781/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TRPV1 antagonist

A 425619 is a potent TRPV1 antagonist (IC50 = 3-9 nM). Blocks capsaicin-evoked currents in rat DRG neurons and inhibits TRPV1-activation by anandamide (Cat. No. 1339) and N-arachidonoyl-dopamine. Reduces pain in inflammatory, postoperative and osteoarthritic in vivo pain models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-5-Isoquinolinyl-N′-[[(4-(trifluoromethyl)phenyl]methyl]urea
CAS 581809-67-8
Quantity 10 mg
Target TRPV Antagonists
Molecular Formula C18H14F3N3O
Formula Weight 345.32
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.