missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 485

Description
A 485 is a potent and selective p300/CREB-binding protein (CBP) HAT domain inhibitor (IC50 values are 2.6 and 9.8 nM for the CBP-bromodomain HAT-C/H3 (BHC) and p300-BHC domains, respectively), which displays > 1000-fold selectivity over closely related HATs. A 485 suppresses proliferation in several hematological malignancies and AR+ prostate cancer cell lines in vitro, and also inhibits tumor growth in a castration-resistant prostate cancer xenograft model. A 485 is orally bioavailable.
To request the negative control for A 485, please fill out the A 486 request form on the SGC website.
Specifications
Specifications
| CAS | 1889279-16-6 |
| Synonym | (1R)-N-[(4-Fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide |
| Quantity | 5 mg |
| Molecular Formula | C25H24F4N4O5 |
| Formula Weight | 536.48 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?