missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ A 83-01

Product Code. 18740597
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18740597 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18740597 Supplier R&D Systems™ Supplier No. TB2939RMU/10
  • ONLINE EXCLUSIVE PRICE
    654.79 €
    590.10 € / 10mg
    Save 64.69 €
    9% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. A 83-01 synthesized to Ancillary Material Grade

TB2939-RMU is an Ancillary Material Grade version of A 83-01 (Cat. No. 2939). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Find out more about our Ancillary Material Grade product range.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(6-Methyl-2-pyridinyl)-N-phenyl-4-(4-quinolinyl)-1 H-pyrazole-1-carbothioamide
CAS 909910-43-6
Quantity 10 mg
Product Line Tocris
Target Compounds for Stem Cell Differentiation
Molecular Formula C25H19N5S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.