missing translation for 'onlineSavingsMsg'
Learn More

A 83-01, TRC

Product Code. 30138123
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138123 10 mg 10mg
30136129 50 mg 50mg
17977394 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138123 Supplier Toronto Research Chemicals Supplier No. TRCA10121010MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 909910-43-6
Molecular Formula C25 H19 N5 S
Molecular Weight (g/mol) 421.52
Synonym A 83-01, 3-(6-Methyl-2-pyridinyl)-N-phenyl-4-(4-quinolinyl)-1H-pyrazole-1-carbothioamide
IUPAC Name 3-(6-methyl-2-pyridyl)-N-phenyl-4-(4-quinolyl)pyrazole-1-carbothioamide
SMILES Cc1cccc(n1)c2nn(cc2c3ccnc4ccccc34)C(=S)Nc5ccccc5

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H19N5S/c1-17-8-7-13-23(27-17)24-21(19-14-15-26-22-12-6-5-11-20(19)22)16-30(29-24)25(31)28-18-9-3-2-4-10-18/h2-16H,1H3,(H,28,31)
Formula Weight 421.1361
Chemical Name or Material 3-(6-Methyl-2-pyridinyl)-N-phenyl-4-(4-quinolinyl)-1H-pyrazole-1-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.