missing translation for 'onlineSavingsMsg'
Learn More

a,a,a-Trifluorotoluene, TRC

Product Code. 30449645
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449645 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449645 Supplier Toronto Research Chemicals Supplier No. TRCT7935251L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha,alpha,alpha-Trifluorotoluene
CAS 98-08-8
Molecular Formula C7 H5 F3
Quantity 1 L
Synonym Trifluoromethylbenzene, Benzene','(trifluoromethyl)-, Toluene','α,α,α-trifluoro- (6CI,8CI), Toluene','α-trifluoro- (3CI), (Trifluoromethyl)benzene, (1,1',1''-Trifluoromethyl)benzene, 1',1',1'-Trifluorotoluene, Benzenyl fluoride, Benzotrifluoride, Benzylidyne fluoride, NSC 8038, Oxsol 2000, Phenylfluoroform, Trifluoro(phenyl)methane, UN 238, α,α,α-Trifluorotoluene
InChI Formula InChI=1S/C7H5F3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H
SMILES FC(F)(F)c1ccccc1
IUPAC Name trifluoromethylbenzene
Molecular Weight (g/mol) 146.11
Formula Weight 146.0343
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.