missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ a-Cyclodextrin

Product Code. 18744374
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18744374 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18744374 Supplier R&D Systems™ Supplier No. 3180/1G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cyclic complexing agent

α-Cyclodextrin (α-CD) is a cylindrical oligosaccharide presenting a hydrophilic surface and a lipophilic cavity. α-Cyclodextrin can form inclusion complexes with a diverse range of hydrophobic compounds. These complexes increase the aqueous solubility, dissolution rate, and bioavailability of the guest compounds.
TRUSTED_SUSTAINABILITY

Specifications

CAS 10016-20-3
Quantity 1 g
Product Line Tocris
Target General Laboratory Research Reagent
Molecular Formula C36H60O30
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.